missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Dog (Beagle) Microsomes
Description
Liver microsomes prepared from a pool of male Beagle Dogs.
Total protein concentration: 20 mg/ml
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
Related Links:
Specifications
Specifications
| Concentration | 20 mg/mL |
| Product Type | Subcellular Fractions |
| Content And Storage | Store between -60°C and -85°C |
| Donor Source | Pooled |
| Gender | Male |
| Shipping Condition | Dry Ice |
| Sub Cellular Localization | Microsomes |
| Target Species | Dog/Canine |
| Tissue | Liver |
| Form | Frozen |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.