Learn More
Dofetilide, Thermo Scientific™
Specifications
| CAS | 115256-11-6 |
|---|---|
| Quantity | 1 g |
| Solubility | Solubility aq. soln.: .. Other solubilities: soluble in acetone, slightly soluble in alcohol |
| Melting Point | 149.0°C to 153.0°C |
| Chemical Name or Material | Dofetilide |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC463370010
|
Thermo Scientific Chemicals
463370010 |
1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC463372500
|
Thermo Scientific Chemicals
463372500 |
250 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 115256-11-6 | |
| Solubility aq. soln.: .. Other solubilities: soluble in acetone, slightly soluble in alcohol | |
| Dofetilide |
| 1 g | |
| 149.0°C to 153.0°C |
Safety and Handling
GHS H Statement Harmful if swallowed.
May damage the unborn child.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
RUO â Research Use Only