Learn More
Dofetilide, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 463372500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Dofetilide | |
| 250 mg | |
| 149.0°C to 153.0°C |
| 115256-11-6 | |
| Solubility aq. soln.: .. Other solubilities: soluble in acetone, slightly soluble in alcohol |
Safety and Handling
GHS H Statement Harmful if swallowed.
May damage the unborn child.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
RUO â Research Use Only