missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Dofetilide
Description
Dofetilide is a selective potassium channel blocker. Blocks KV11.1 (hERG) channels; inhibits the rapid delayed-rectifier K+ current (IKr). Displays class III antiarrhythmic properties.
Specifications
Specifications
| Chemical Name or Material | 1-(4-Methanesulphonamidophenoxy)-2-[N-(4-methanesulphonamidophenethyl)-N-methylamino]ethane |
| CAS | 115256-11-6 |
| Assay Percent Range | >99% |
| Quantity | 25 mg |
| Molecular Formula | C19H27N3O5S2 |
| Formula Weight | Observed MW: 441.56 |