missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecylbenzene Sodium Sulfonate, Powder, Approx. 90%, Spectrum™ Chemical
D1064, 25155-30-0, C18H29NaO3S
$196.14 - $7595.17
Chemical Identifiers
| CAS | 25155-30-0 |
|---|---|
| Molecular Formula | C18H29NaO3S |
| Molecular Weight (g/mol) | 348.48 |
| InChI Key | HFQQZARZPUDIFP-UHFFFAOYSA-M |
| IUPAC Name | sodium 2-dodecylbenzene-1-sulfonate |
| SMILES | [Na+].CCCCCCCCCCCCC1=C(C=CC=C1)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602315
|
Spectrum Chemical Mfg Cor
D1064100GM |
100 g | Poly Bottle |
Each for $196.14
|
|
||||
|
18602316
|
Spectrum Chemical Mfg Cor
D1064500GM |
500 g | Poly Bottle |
Each for $253.71
|
|
||||
|
18602317
|
Spectrum Chemical Mfg Cor
D10643KG |
3 kg | Poly Pail |
Each for $861.57
|
|
||||
|
18602318
|
Spectrum Chemical Mfg Cor
D106412KG |
12 kg | Poly Pail |
Each for $1,383.37
|
|
||||
|
18602319
|
Spectrum Chemical Mfg Cor
D106425KGBL |
25 kg | Fiber Drum |
Each for $2,710.80
|
|
||||
|
18602320
|
Spectrum Chemical Mfg Cor
D106490KGBL |
90 kg | Fiber Drum |
Each for $7,595.17
|
|
||||
Description
Spectrum™ Chemical Dodecylbenzene Sodium Sulfonate, PowderChemical Identifiers
| 25155-30-0 | |
| 348.48 | |
| sodium 2-dodecylbenzene-1-sulfonate |
| C18H29NaO3S | |
| HFQQZARZPUDIFP-UHFFFAOYSA-M | |
| [Na+].CCCCCCCCCCCCC1=C(C=CC=C1)S([O-])(=O)=O |
Specifications
| 25155-30-0 | |
| Poly Bottle | |
| 100 g | |
| [Na+].CCCCCCCCCCCCC1=C(C=CC=C1)S([O-])(=O)=O | |
| 348.48 | |
| Reagent |
| 100% | |
| C18H29NaO3S | |
| HFQQZARZPUDIFP-UHFFFAOYSA-M | |
| sodium 2-dodecylbenzene-1-sulfonate | |
| ∼90% |