missing translation for 'onlineSavingsMsg'
Learn More

Dodecylbenzene Sodium Sulfonate, Powder, Approx. 90%, Spectrum™ Chemical

Catalog No. 18602315
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
12 kg
25 kg
90 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602315 100 g Poly Bottle
18602316 500 g Poly Bottle
18602317 3 kg Poly Pail
18602318 12 kg Poly Pail
18602319 25 kg Fiber Drum
18602320 90 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18602315 Supplier Spectrum Chemical Mfg Cor Supplier No. D1064100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

D1064, 25155-30-0, C18H29NaO3S

Spectrum™ Chemical Dodecylbenzene Sodium Sulfonate, Powder

Chemical Identifiers

CAS 25155-30-0
Molecular Formula C18H29NaO3S
Molecular Weight (g/mol) 348.48
InChI Key HFQQZARZPUDIFP-UHFFFAOYSA-M
IUPAC Name sodium 2-dodecylbenzene-1-sulfonate
SMILES [Na+].CCCCCCCCCCCCC1=C(C=CC=C1)S([O-])(=O)=O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 100 g
Percent Purity ∼90%
Grade Reagent