missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecylbenzene Sodium Sulfonate, Powder, Approx. 90%, Spectrum™ Chemical
D1064, 25155-30-0, C18H29NaO3S
Supplier: Spectrum Chemical Mfg Cor D10643KG
Description
Spectrum™ Chemical Dodecylbenzene Sodium Sulfonate, PowderSpecifications
| 25155-30-0 | |
| Poly Pail | |
| 3 kg | |
| [Na+].CCCCCCCCCCCCC1=C(C=CC=C1)S([O-])(=O)=O | |
| 348.48 | |
| Reagent |
| 100% | |
| C18H29NaO3S | |
| HFQQZARZPUDIFP-UHFFFAOYSA-M | |
| sodium 2-dodecylbenzene-1-sulfonate | |
| ∼90% |
Chemical Identifiers
| 25155-30-0 | |
| 348.48 | |
| sodium 2-dodecylbenzene-1-sulfonate |
| C18H29NaO3S | |
| HFQQZARZPUDIFP-UHFFFAOYSA-M | |
| [Na+].CCCCCCCCCCCCC1=C(C=CC=C1)S([O-])(=O)=O |