missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyl sulfate, sodium salt, 85%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White to Yellow |
| Physical Form | Powder or Crystals |
| Melting Point | 206.0°C |
| Quantity | 100 g |
| Flash Point | >150°C |
| Infrared Spectrum | Authentic |
| Packaging | Plastic Bottle |
| Assay Percent Range | 85% |
| Linear Formula | CH3(CH2)11OSO3Na |
| Solubility Information | Solubility in water: 130g/L (20°C). |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye damage. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. |
RUO – Research Use Only