missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl sulfate, sodium salt, 85%, pure

Catalog No. AC226141000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226141000 100 g Plastic Bottle
AC226145000 500 g Plastic Bottle
AC226140025 2.5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226141000 Supplier Thermo Scientific Chemicals Supplier No. 226141000

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 151-21-3 , 7757-82-6 , 7647-14-5
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White to Yellow
Physical Form Powder or Crystals
Melting Point 206.0°C
Quantity 100 g
Flash Point >150°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Assay Percent Range 85%
Linear Formula CH3(CH2)11OSO3Na
Solubility Information Solubility in water: 130g/L (20°C).
Formula Weight 288.38
Percent Purity 85%
Grade Pure
Chemical Name or Material Dodecyl sulfate, sodium salt
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only