missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Methyl Sulfide 98.0+%, TCI America™

Catalog No. p-7127902
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37675G 5 g
D376725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37675G Supplier TCI America Supplier No. D37675G
Only null left

Chemical Identifiers

CAS 3698-89-3
Molecular Formula C13H28S
Molecular Weight (g/mol) 216.43
MDL Number MFCD00015073
InChI Key KJWHJDGMOQJLGF-UHFFFAOYSA-N
Synonym dodecyl methyl sulfide, 2-thiatetradecane, 1-methylsulfanyl dodecane, methylthiododecane, methyl lauryl sulfide, dodecane, 1-methylthio, methyl n-dodecyl sulfide, dodecyl methyl sulphide, sulfide, dodecyl methyl, unii-77bo1dv08u
PubChem CID 77287
IUPAC Name 1-(methylsulfanyl)dodecane
SMILES CCCCCCCCCCCCSC

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 216.43
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecyl Methyl Sulfide