missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyl Methacrylate (stabilized with MEHQ) 97.0+%, TCI America™
$41.77 - $125.31
Chemical Identifiers
| CAS | 142-90-5 |
|---|---|
| Molecular Formula | C16H30O2 |
| Molecular Weight (g/mol) | 254.414 |
| MDL Number | MFCD00008972 |
| InChI Key | GMSCBRSQMRDRCD-UHFFFAOYSA-N |
| Synonym | dodecyl methacrylate, lauryl methacrylate, dodecyl 2-methylacrylate, metazene, n-dodecyl methacrylate, sipomer lma, 2-propenoic acid, 2-methyl-, dodecyl ester, dodecyl 2-methyl-2-propenoate, lama, ageflex fm 246 |
| PubChem CID | 8906 |
| IUPAC Name | dodecyl 2-methylprop-2-enoate |
| SMILES | CCCCCCCCCCCCOC(=O)C(=C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
M008325ML
|
N/A | 25 mL |
Each for $41.77
|
|
|||||
|
M0083100ML
|
N/A | 100 mL |
Each for $60.75
|
|
|||||
|
M0083500ML
|
N/A | 500 mL |
Each for $125.31
|
|
|||||
Chemical Identifiers
| C16H30O2 | |
| MFCD00008972 | |
| dodecyl methacrylate, lauryl methacrylate, dodecyl 2-methylacrylate, metazene, n-dodecyl methacrylate, sipomer lma, 2-propenoic acid, 2-methyl-, dodecyl ester, dodecyl 2-methyl-2-propenoate, lama, ageflex fm 246 | |
| dodecyl 2-methylprop-2-enoate |
Specifications
| 142-90-5 | |
| 167°C | |
| MFCD00008972 | |
| dodecyl methacrylate, lauryl methacrylate, dodecyl 2-methylacrylate, metazene, n-dodecyl methacrylate, sipomer lma, 2-propenoic acid, 2-methyl-, dodecyl ester, dodecyl 2-methyl-2-propenoate, lama, ageflex fm 246 | |
| CCCCCCCCCCCCOC(=O)C(=C)C | |
| 254.414 | |
| 254.41 | |
| Liquid |
| Colorless | |
| C16H30O2 | |
| 3082 | |
| GMSCBRSQMRDRCD-UHFFFAOYSA-N | |
| dodecyl 2-methylprop-2-enoate | |
| 8906 | |
| ≥97.0% (GC) | |
| Dodecyl Methacrylate (stabilized with MEHQ) |
Safety and Handling
EINECSNumber : (2)-1039
RTECSNumber : OZ4300000
TSCA : Yes