missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Methacrylate (stabilized with MEHQ) 97.0+%, TCI America™

Supplier:  TCI America M0083100ML

Quantity 100 mL

 View more versions of this product

Catalog No. M0083100ML


Only null left

Description

Description

Specifications

Chemical Identifiers

142-90-5
254.414
GMSCBRSQMRDRCD-UHFFFAOYSA-N
8906
CCCCCCCCCCCCOC(=O)C(=C)C
C16H30O2
MFCD00008972
dodecyl methacrylate, lauryl methacrylate, dodecyl 2-methylacrylate, metazene, n-dodecyl methacrylate, sipomer lma, 2-propenoic acid, 2-methyl-, dodecyl ester, dodecyl 2-methyl-2-propenoate, lama, ageflex fm 246
dodecyl 2-methylprop-2-enoate

Specifications

Dodecyl Methacrylate (stabilized with MEHQ)
Colorless
C16H30O2
MFCD00008972
dodecyl methacrylate, lauryl methacrylate, dodecyl 2-methylacrylate, metazene, n-dodecyl methacrylate, sipomer lma, 2-propenoic acid, 2-methyl-, dodecyl ester, dodecyl 2-methyl-2-propenoate, lama, ageflex fm 246
CCCCCCCCCCCCOC(=O)C(=C)C
254.414
254.41
Liquid
142-90-5
167°C
100 mL
3082
GMSCBRSQMRDRCD-UHFFFAOYSA-N
dodecyl 2-methylprop-2-enoate
8906
≥97.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1039

RTECSNumber : OZ4300000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions