missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Methacrylate (stabilized with MEHQ) 97.0+%, TCI America™

Catalog No. M008325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M008325ML 25 mL
M0083100ML 100 mL
M0083500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M008325ML Supplier TCI America Supplier No. M008325ML

Chemical Identifiers

CAS 142-90-5
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.414
MDL Number MFCD00008972
InChI Key GMSCBRSQMRDRCD-UHFFFAOYSA-N
Synonym dodecyl methacrylate, lauryl methacrylate, dodecyl 2-methylacrylate, metazene, n-dodecyl methacrylate, sipomer lma, 2-propenoic acid, 2-methyl-, dodecyl ester, dodecyl 2-methyl-2-propenoate, lama, ageflex fm 246
PubChem CID 8906
IUPAC Name dodecyl 2-methylprop-2-enoate
SMILES CCCCCCCCCCCCOC(=O)C(=C)C

Specifications

Color Colorless
Boiling Point 167°C
Quantity 25 mL
UN Number 3082
Formula Weight 254.41
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1039
RTECSNumber OZ4300000
TSCA Yes