missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™
Supplier: TCI America D4129100ML
| Quantity | 100 mL |
|---|
Chemical Identifiers
| C15H28O2 | |
| MFCD00042871 | |
| dodecyl acrylate, lauryl acrylate, n-lauryl acrylate, n-dodecyl acrylate, 2-propenoic acid, dodecyl ester, acrylic acid, dodecyl ester, acrylic acid lauryl ester, acrylic acid dodecyl ester, unii-90ik36wgvr, 90ik36wgvr | |
| dodecyl prop-2-enoate |
Specifications
| Dodecyl Acrylate (stabilized with MEHQ) | |
| 4°C | |
| 120°C | |
| 100 mL | |
| dodecyl acrylate, lauryl acrylate, n-lauryl acrylate, n-dodecyl acrylate, 2-propenoic acid, dodecyl ester, acrylic acid, dodecyl ester, acrylic acid lauryl ester, acrylic acid dodecyl ester, unii-90ik36wgvr, 90ik36wgvr | |
| CCCCCCCCCCCCOC(=O)C=C | |
| 240.39 | |
| 240.39 | |
| Liquid |
| 2156-97-0 | |
| Colorless | |
| C15H28O2 | |
| MFCD00042871 | |
| PBOSTUDLECTMNL-UHFFFAOYSA-N | |
| dodecyl prop-2-enoate | |
| 75084 | |
| ≥98.0% (GC) |
Safety and Handling
EINECSNumber : (2)-0990
TSCA : Yes