missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. p-7128116
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D412925ML 25 mL
D4129100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D412925ML Supplier TCI America Supplier No. D412925ML

Chemical Identifiers

CAS 2156-97-0
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.39
MDL Number MFCD00042871
InChI Key PBOSTUDLECTMNL-UHFFFAOYSA-N
Synonym dodecyl acrylate, lauryl acrylate, n-lauryl acrylate, n-dodecyl acrylate, 2-propenoic acid, dodecyl ester, acrylic acid, dodecyl ester, acrylic acid lauryl ester, acrylic acid dodecyl ester, unii-90ik36wgvr, 90ik36wgvr
PubChem CID 75084
IUPAC Name dodecyl prop-2-enoate
SMILES CCCCCCCCCCCCOC(=O)C=C

Specifications

Melting Point 4°C
Color Colorless
Boiling Point 120°C
Quantity 25 mL
Formula Weight 240.39
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0990
TSCA Yes