missing translation for 'onlineSavingsMsg'
Learn More

Dodecanedioic Acid 99.0+%, TCI America™

Supplier:  TCI America D001325G

 View more versions of this product

Catalog No. D001325G


Only null left

Description

Description

Specifications

Specifications

Dodecanedioic Acid
129°C
C12H22O4
25 g
TVIDDXQYHWJXFK-UHFFFAOYSA-N
dodecanedioic acid
12736
230.30
Crystalline Powder
693-23-2
White
MFCD00002735
1,12-dodecanedioic acid, 1,10-decanedicarboxylic acid, decamethylenedicarboxylic acid, 1,10-dicarboxydecane, n-dodecanedioic acid, unii-978yu42q6i, dsstox_cid_7297, dsstox_rid_78394, dsstox_gsid_27297, n-dodecanedioate
C(CCCCCC(=O)O)CCCCC(=O)O
230.304
CHEBI:4676
≥99.0% (GC)

Chemical Identifiers

693-23-2
230.304
TVIDDXQYHWJXFK-UHFFFAOYSA-N
12736
dodecanedioic acid
C12H22O4
MFCD00002735
1,12-dodecanedioic acid, 1,10-decanedicarboxylic acid, decamethylenedicarboxylic acid, 1,10-dicarboxydecane, n-dodecanedioic acid, unii-978yu42q6i, dsstox_cid_7297, dsstox_rid_78394, dsstox_gsid_27297, n-dodecanedioate
CHEBI:4676
C(CCCCCC(=O)O)CCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0878

RTECSNumber : JR2370000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions