missing translation for 'onlineSavingsMsg'
Learn More

Dodecanedioic Acid 99.0+%, TCI America™

Catalog No. p-7125982
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D001325G 25 g
D0013500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D001325G Supplier TCI America Supplier No. D001325G
Only null left

Chemical Identifiers

CAS 693-23-2
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00002735
InChI Key TVIDDXQYHWJXFK-UHFFFAOYSA-N
Synonym 1,12-dodecanedioic acid, 1,10-decanedicarboxylic acid, decamethylenedicarboxylic acid, 1,10-dicarboxydecane, n-dodecanedioic acid, unii-978yu42q6i, dsstox_cid_7297, dsstox_rid_78394, dsstox_gsid_27297, n-dodecanedioate
PubChem CID 12736
ChEBI CHEBI:4676
IUPAC Name dodecanedioic acid
SMILES C(CCCCCC(=O)O)CCCCC(=O)O

Specifications

Melting Point 129°C
Color White
Quantity 25 g
Formula Weight 230.30
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dodecanedioic Acid
EINECSNumber (2)-0878
RTECSNumber JR2370000
TSCA Yes