accessibility menu, dialog, popup

UserName

HOOOHO

CAS RN 693-23-2

IUPAC Name: dodecanedioic acid
Synonyms: dodecanedioic acid n-dodecane-a,w-dioate SL-AH corfree M 2 sodium salt dodecanedioic acid n-dodecane-a,w-dioic acid
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: TVIDDXQYHWJXFK-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)