missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecamethylcyclohexasiloxane 97.0+%, TCI America™
$146.31 - $1341.85
Chemical Identifiers
| CAS | 540-97-6 |
|---|---|
| Molecular Formula | C12H36O6Si6 |
| Molecular Weight (g/mol) | 444.924 |
| MDL Number | MFCD00144215 |
| InChI Key | IUMSDRXLFWAGNT-UHFFFAOYSA-N |
| PubChem CID | 10911 |
| IUPAC Name | 2,2,4,4,6,6,8,8,10,10,12,12-dodecamethyl-1,3,5,7,9,11-hexaoxa-2,4,6,8,10,12-hexasilacyclododecane |
| SMILES | C[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)C |
Chemical Identifiers
| 540-97-6 | |
| 444.924 | |
| IUMSDRXLFWAGNT-UHFFFAOYSA-N | |
| 2,2,4,4,6,6,8,8,10,10,12,12-dodecamethyl-1,3,5,7,9,11-hexaoxa-2,4,6,8,10,12-hexasilacyclododecane |
| C12H36O6Si6 | |
| MFCD00144215 | |
| 10911 | |
| C[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)C |
Specifications
| 540-97-6 | |
| 245°C | |
| MFCD00144215 | |
| IUMSDRXLFWAGNT-UHFFFAOYSA-N | |
| 2,2,4,4,6,6,8,8,10,10,12,12-dodecamethyl-1,3,5,7,9,11-hexaoxa-2,4,6,8,10,12-hexasilacyclododecane | |
| 10911 | |
| ≥97.0% (GC) | |
| Dodecamethylcyclohexasiloxane |
| Colorless | |
| C12H36O6Si6 | |
| 1 mL | |
| C[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)C | |
| 444.924 | |
| 444.92 | |
| Liquid |
Safety and Handling
RTECSNumber : GW2328550
TSCA : Yes