accessibility menu, dialog, popup

UserName

CH3SiH3COSiCH3CH3OSiCH3CH3OSiCH3H3COSiH3CH3COSiH3CH3CO

CAS RN 540-97-6

IUPAC Name: dodecamethylcyclohexasiloxane
Synonyms: dodecamethylcyclohexasiloxane
Molecular Weight (g/mol): 444.92
Molecular Formula: C12H36O6Si6
InChi Key: IUMSDRXLFWAGNT-UHFFFAOYSA-N
SMILES: C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results