missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecamethylcyclohexasiloxane 97.0+%, TCI America™
Supplier: TCI America D20405ML
Specifications
| Dodecamethylcyclohexasiloxane | |
| Colorless | |
| C12H36O6Si6 | |
| 5 mL | |
| C[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)C | |
| 444.924 | |
| 444.92 | |
| Liquid |
| 540-97-6 | |
| 245°C | |
| MFCD00144215 | |
| IUMSDRXLFWAGNT-UHFFFAOYSA-N | |
| 2,2,4,4,6,6,8,8,10,10,12,12-dodecamethyl-1,3,5,7,9,11-hexaoxa-2,4,6,8,10,12-hexasilacyclododecane | |
| 10911 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 540-97-6 | |
| 444.924 | |
| IUMSDRXLFWAGNT-UHFFFAOYSA-N | |
| 2,2,4,4,6,6,8,8,10,10,12,12-dodecamethyl-1,3,5,7,9,11-hexaoxa-2,4,6,8,10,12-hexasilacyclododecane |
| C12H36O6Si6 | |
| MFCD00144215 | |
| 10911 | |
| C[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)C |
Safety and Handling
RTECSNumber : GW2328550
TSCA : Yes