missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Docetaxel

Catalog No. 405610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
405610 10 mg
405650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 405610 Supplier Tocris Bioscience™ Supplier No. 4056/10

Microtubule stabilizer

Binds and stabilizes microtubules; leads to cell cycle arrest and apoptosis. Displays significant antitumor activity in ovarian and gastric tumors. Prevents microtubule depolymerization and disassembly in the absence of GTP. Exhibits cytotoxic activity.

Chemical Identifiers

CAS 114977-28-5
Molecular Formula C43H53NO14

Specifications

Quantity 10 mg
Formula Weight 807.88
Purity >99%
Molecular Weight (g/mol) 807.89
Chemical Name or Material Docetaxel
Recommended Storage Store at +4°C