missing translation for 'onlineSavingsMsg'
Learn More

Dobutamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7128230
Change view
Click to view available options
Quantity:
100 mg
20 mg
Catalog No. Quantity
D433520MG 20 mg
D4335100MG 100 mg
2 options

Catalog No. D433520MG

Supplier: TCI America D433520MG

Only null left

Chemical Identifiers

CAS 49745-95-1
Molecular Formula C18H24ClNO3
Molecular Weight (g/mol) 337.84
MDL Number MFCD00153795
InChI Key BQKADKWNRWCIJL-UHFFFAOYNA-N
Synonym dobutamine hydrochloride, dobutamine hcl, dobutrex, inotrex, dl-dobutamine hydrochloride, dobutamina clorhidrato spanish, 4-2-4-4-hydroxyphenyl butan-2-ylamino ethyl benzene-1,2-diol hydrochloride, +--4-2-3-p-hydroxyphenyl-1-methylpropyl amino ethyl pyrocatechol hydrochloride, 1,2-benzenediol, 4-2-3-4-hydroxyphenyl-1-methylpropyl amino ethyl-, hydrochloride, +-
PubChem CID 65324
ChEBI CHEBI:4671
IUPAC Name hydrogen 4-(2-{[4-(4-hydroxyphenyl)butan-2-yl]amino}ethyl)benzene-1,2-diol chloride
SMILES [H+].[Cl-].CC(CCC1=CC=C(O)C=C1)NCCC1=CC=C(O)C(O)=C1

Specifications

Melting Point 189°C
Quantity 20 mg
Formula Weight 337.84
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dobutamine Hydrochloride