accessibility menu, dialog, popup

UserName

HClCH3HOHNOHOH

CAS RN 49745-95-1

IUPAC Name: hydrogen 4-(2-{[4-(4-hydroxyphenyl)butan-2-yl]amino}ethyl)benzene-1,2-diol chloride
Synonyms: dobutamine hydrochloride [usan:ban:jan]
Molecular Weight (g/mol): 337.84
Molecular Formula: C18H24ClNO3
InChi Key: BQKADKWNRWCIJL-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(CCC1=CC=C(O)C=C1)NCCC1=CC=C(O)C(O)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results