missing translation for 'onlineSavingsMsg'
Learn More

Dobutamine Hydrochloride 98.0+%, TCI America™

Catalog No. D433520MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D433520MG 20 mg
D4335100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D433520MG Supplier TCI America Supplier No. D433520MG

Chemical Identifiers

CAS 49745-95-1
Molecular Formula C18H24ClNO3
Molecular Weight (g/mol) 337.84
MDL Number MFCD00153795
InChI Key BQKADKWNRWCIJL-UHFFFAOYNA-N
Synonym dobutamine hydrochloride, dobutamine hcl, dobutrex, inotrex, dl-dobutamine hydrochloride, dobutamina clorhidrato spanish, 4-2-4-4-hydroxyphenyl butan-2-ylamino ethyl benzene-1,2-diol hydrochloride, +--4-2-3-p-hydroxyphenyl-1-methylpropyl amino ethyl pyrocatechol hydrochloride, 1,2-benzenediol, 4-2-3-4-hydroxyphenyl-1-methylpropyl amino ethyl-, hydrochloride, +-
PubChem CID 65324
ChEBI CHEBI:4671
IUPAC Name hydrogen 4-(2-{[4-(4-hydroxyphenyl)butan-2-yl]amino}ethyl)benzene-1,2-diol chloride
SMILES [H+].[Cl-].CC(CCC1=CC=C(O)C=C1)NCCC1=CC=C(O)C(O)=C1

Specifications

Melting Point 189°C
Quantity 20 mg
Formula Weight 337.84
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dobutamine Hydrochloride