missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DMH-1

Catalog No. 412650
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
412650 50 mg
412610 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 412650 Supplier Tocris Bioscience™ Supplier No. 4126/50
Only null left

Selective ALK2 inhibitor

DMH-1 is a selective inhibitor of bone morphogenic protein (BMP) type-I receptor activin receptor-like kinase 2 (ALK2) receptor (IC50 = 108 nM or 12.6 nM in in vitro kinase assays). DMH-1 exhibits 6- and 19-fold selectivity for ALK-2 over ALK-1 and ALK-3, respectively, and no significant inhibition of AMPK, ALK5, KDR (VEGFR-2) or PDGFRβ receptors. DMH-1 blocks BMP4-induced phosphorylation of Smads 1, 5 and 8 in HEK293 cells. Promotes neurogenesis in human induced pluripotent stem cells (iPSCs) when used in combination with SB 431542 (Cat. No. 1614). DMH-1 suppresses lung cancer cell proliferation, migration, invasion in vitro and reduces tumor growth in a mouse lung cancer xenograft model. DMH-1 inhibits cellular autophagy responses. DMH-1 induces intestinal differentiation in human intestinal organoids (hIOs) derived from human pluripotent stems cells (hPSCs).

Chemical Identifiers

CAS 1206711-16-1
Molecular Formula C24H20N4O

Specifications

Quantity 50 mg
Formula Weight 380.44
Purity >98%
Molecular Weight (g/mol) 380.45
Chemical Name or Material DMH-1
Recommended Storage Store at +4°C