missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DLG5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5122033

 View more versions of this product

Catalog No. PIPA5122033


Only null left
Explore available promotions

Description

Description

May play a role at the plasma membrane in the maintenance of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.
Specifications

Specifications

DLG5
Polyclonal
Unconjugated
Dlg5
4933429D20Rik; discs large homolog 5; discs large MAGUK scaffold protein 5; discs large protein LP-DLG; discs large protein P-dlg; discs, large homolog 5; discs, large homolog 5 (Drosophila); disks large homolog 5; DLG5; KIAA0583; large type of P-DLG; LP-DLG; mKIAA0583; PDLG; P-DLG5; Placenta and prostate DLG; T25557
Rabbit
Antigen affinity chromatography, Protein A
RUO
9231
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q8TDM6
Dlg5
Asp1722-Asn1905 (Accession Q8TDM6), with N-terminal His Tag.
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions