missing translation for 'onlineSavingsMsg'
Learn More

DLG5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

$323.41 - $787.99

Specifications

Antigen DLG5
Concentration 0.5 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PIPA5122033
View Documents
Invitrogen™
PA5122033
20 μL
Each for $323.41
Only null left
 
PIPA5122032
View Documents
Invitrogen™
PA5122032
100 μL
Each for $787.99
Only null left
 
Description

Description

May play a role at the plasma membrane in the maintenance of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.
Specifications

Specifications

DLG5
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
Q8TDM6
9231
Asp1722-Asn1905 (Accession Q8TDM6), with N-terminal His Tag.
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Dlg5
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
4933429D20Rik; discs large homolog 5; discs large MAGUK scaffold protein 5; discs large protein LP-DLG; discs large protein P-dlg; discs, large homolog 5; discs, large homolog 5 (Drosophila); disks large homolog 5; DLG5; KIAA0583; large type of P-DLG; LP-DLG; mKIAA0583; PDLG; P-DLG5; Placenta and prostate DLG; T25557
Dlg5
IgG
Antigen affinity chromatography, Protein A
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'