Learn More
Thermo Scientific Chemicals DL-Tyrosine, 98%
CAS: 556-03-6 | C9H11NO3 | 181.19 g/mol
$61.81 - $759.05
Chemical Identifiers
| CAS | 556-03-6 |
|---|---|
| Molecular Formula | C9H11NO3 |
| Molecular Weight (g/mol) | 181.19 |
| MDL Number | MFCD00063074 |
| InChI Key | OUYCCCASQSFEME-UHFFFAOYNA-N |
| Synonym | dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s |
| PubChem CID | 1153 |
| ChEBI | CHEBI:18186 |
| IUPAC Name | 2-amino-3-(4-hydroxyphenyl)propanoic acid |
| SMILES | NC(CC1=CC=C(O)C=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1374009
|
Thermo Scientific Chemicals
A1374009 |
10 g |
Each for $61.81
|
|
|||||
|
AAA1374018
|
Thermo Scientific Chemicals
A1374018 |
50 g |
Each for $165.40
|
|
|||||
|
AAA1374030
|
Thermo Scientific Chemicals
A1374030 |
250 g |
Each for $759.05
|
|
|||||
Description
The tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 556-03-6 | |
| 181.19 | |
| OUYCCCASQSFEME-UHFFFAOYNA-N | |
| 1153 | |
| 2-amino-3-(4-hydroxyphenyl)propanoic acid |
| C9H11NO3 | |
| MFCD00063074 | |
| dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s | |
| CHEBI:18186 | |
| NC(CC1=CC=C(O)C=C1)C(O)=O |
Specifications
| ∼325°C (decomposition) | |
| C9H11NO3 | |
| 515881 | |
| dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s | |
| OUYCCCASQSFEME-UHFFFAOYNA-N | |
| 2-amino-3-(4-hydroxyphenyl)propanoic acid | |
| 181.19 | |
| CHEBI:18186 | |
| 98% |
| 556-03-6 | |
| MFCD00063074 | |
| 14,9839 | |
| Soluble in water. | |
| NC(CC1=CC=C(O)C=C1)C(O)=O | |
| 10 g | |
| 1153 | |
| 181.19 | |
| DL-Tyrosine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-113-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only