Learn More
Thermo Scientific Chemicals DL-Tyrosine, 98%
CAS: 556-03-6 | C9H11NO3 | 181.19 g/mol
Supplier: Thermo Scientific Chemicals A1374030
Description
The tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe tyrosine of this diet was labeled by adding to it an amount of synthetic, isotopic dl-tyrosine cor- responding to twice that already present in the food. Incorporation of total C14 into muscle protein was reduced in 186-day infected mice fed dl-tyrosine-2-C-14 as compared to muscle proteins obtained from noninfected animals.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| ∼325°C (decomposition) | |
| 556-03-6 | |
| MFCD00063074 | |
| 14,9839 | |
| Soluble in water. | |
| NC(CC1=CC=C(O)C=C1)C(O)=O | |
| 250 g | |
| 1153 | |
| 181.19 |
| DL-Tyrosine | |
| C9H11NO3 | |
| 515881 | |
| dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s | |
| OUYCCCASQSFEME-UHFFFAOYNA-N | |
| 2-amino-3-(4-hydroxyphenyl)propanoic acid | |
| 181.19 | |
| CHEBI:18186 | |
| 98% |
Chemical Identifiers
| 556-03-6 | |
| 181.19 | |
| OUYCCCASQSFEME-UHFFFAOYNA-N | |
| 1153 | |
| 2-amino-3-(4-hydroxyphenyl)propanoic acid |
| C9H11NO3 | |
| MFCD00063074 | |
| dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s | |
| CHEBI:18186 | |
| NC(CC1=CC=C(O)C=C1)C(O)=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-113-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only