Learn More
Thermo Scientific Chemicals DL-Threonine, 99%
CAS: 80-68-2 | C4H9NO3 | 119.12 g/mol
$81.73 - $872.05
Chemical Identifiers
| CAS | 80-68-2 |
|---|---|
| Molecular Formula | C4H9NO3 |
| Molecular Weight (g/mol) | 119.12 |
| MDL Number | MFCD00063722 |
| InChI Key | AYFVYJQAPQTCCC-PWNYCUMCSA-N |
| Synonym | dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l |
| PubChem CID | 205 |
| ChEBI | CHEBI:38263 |
| SMILES | C[C@@H](O)[C@@H](N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1060614
|
Thermo Scientific Chemicals
A1060614 |
25 g |
Each for $81.73
|
|
|||||
|
AAA1060622
|
Thermo Scientific Chemicals
A1060622 |
100 g |
Each for $210.07
|
|
|||||
|
AAA1060636
|
Thermo Scientific Chemicals
A1060636 |
500 g |
Each for $872.05
|
|
|||||
Description
DL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 80-68-2 | |
| 119.12 | |
| AYFVYJQAPQTCCC-PWNYCUMCSA-N | |
| 205 | |
| C[C@@H](O)[C@@H](N)C(O)=O |
| C4H9NO3 | |
| MFCD00063722 | |
| dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l | |
| CHEBI:38263 |
Specifications
| ∼244°C (decomposition) | |
| C4H9NO3 | |
| 1721647 | |
| dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l | |
| AYFVYJQAPQTCCC-PWNYCUMCSA-N | |
| 2-amino-3-hydroxybutanoic acid | |
| 119.12 | |
| CHEBI:38263 | |
| 99% |
| 80-68-2 | |
| MFCD00063722 | |
| 14,9380 | |
| Soluble in water. | |
| C[C@@H](O)[C@@H](N)C(O)=O | |
| 25 g | |
| 205 | |
| 119.12 | |
| DL-Threonine |
Safety and Handling
EINECSNumber : 201-300-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only