Learn More
Thermo Scientific Chemicals DL-Threonine, 99%
CAS: 80-68-2 | C4H9NO3 | 119.12 g/mol
Supplier: Thermo Scientific Chemicals A1060614
Description
DL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| ∼244°C (decomposition) | |
| 80-68-2 | |
| MFCD00063722 | |
| 14,9380 | |
| Soluble in water. | |
| C[C@@H](O)[C@@H](N)C(O)=O | |
| 25 g | |
| 205 | |
| 119.12 |
| DL-Threonine | |
| C4H9NO3 | |
| 1721647 | |
| dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l | |
| AYFVYJQAPQTCCC-PWNYCUMCSA-N | |
| 2-amino-3-hydroxybutanoic acid | |
| 119.12 | |
| CHEBI:38263 | |
| 99% |
Chemical Identifiers
| 80-68-2 | |
| 119.12 | |
| AYFVYJQAPQTCCC-PWNYCUMCSA-N | |
| 205 | |
| C[C@@H](O)[C@@H](N)C(O)=O |
| C4H9NO3 | |
| MFCD00063722 | |
| dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l | |
| CHEBI:38263 |
Safety and Handling
EINECSNumber : 201-300-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only