missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Threonine, 99%

CAS: 80-68-2 | C4H9NO3 | 119.12 g/mol

Supplier:  Thermo Scientific Chemicals A1060636

 View more versions of this product

Catalog No. AAA1060636

Only null left
Explore available promotions

Description

Description

DL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

∼244°C (decomposition)
80-68-2
MFCD00063722
14,9380
Soluble in water.
C[C@@H](O)[C@@H](N)C(O)=O
500 g
205
119.12
DL-Threonine
C4H9NO3
1721647
dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l
AYFVYJQAPQTCCC-PWNYCUMCSA-N
(2R,3R)-2-amino-3-hydroxybutanoic acid
119.12
CHEBI:38263
99%

Chemical Identifiers

80-68-2
119.12
AYFVYJQAPQTCCC-PWNYCUMCSA-N
205
C[C@@H](O)[C@@H](N)C(O)=O
C4H9NO3
MFCD00063722
dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l
CHEBI:38263
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-300-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only