missing translation for 'onlineSavingsMsg'
Learn More

DL-Ornithine monohydrochloride, 99%

CAS: 1069-31-4 | C5H13ClN2O2 | 168.62 g/mol

$65.45 - $273.33

Chemical Identifiers

CAS 1069-31-4
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.62
MDL Number MFCD00065398
InChI Key GGTYBZJRPHEQDG-UHFFFAOYNA-N
Synonym dl-ornithine hydrochloride, 2,5-diaminopentanoic acid hydrochloride, dl-ornithine monohydrochloride, dl-ornithinehcl, h-dl-orn-oh.hcl, dl-ornithine hcl, dl-ornithine, hydrochloride, ornithine monohydrochloride, ornithine hydrochloride, ornithine, hydrochloride 1:1
PubChem CID 71598
IUPAC Name hydrogen 2,5-diaminopentanoic acid chloride
SMILES [H+].[Cl-].NCCCC(N)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1817306
View Documents Promotion Details
N/A 5 g
Each for $65.45
Only null left
 
AAA1817314
View Documents Promotion Details
N/A 25 g
Each for $174.10
Only null left
 
AAA1817318
View Documents Promotion Details
N/A 50 g
Each for $273.33
Only null left
 
Description

Description

Erythro-3-hydroxy-DL-ornithine monohydrochloride is used as a reagent to synthesize DL-Capreomycidine, a guanidine (HCl)-containing amino acid that is a component of antituberculous peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erythro-3-hydroxy-DL-ornithine monohydrochloride is used as a reagent to synthesize DL-Capreomycidine, a guanidine (HCl)-containing amino acid that is a component of antituberculous peptides.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

1069-31-4
168.62
GGTYBZJRPHEQDG-UHFFFAOYNA-N
71598
[H+].[Cl-].NCCCC(N)C(O)=O
C5H13ClN2O2
MFCD00065398
dl-ornithine hydrochloride, 2,5-diaminopentanoic acid hydrochloride, dl-ornithine monohydrochloride, dl-ornithinehcl, h-dl-orn-oh.hcl, dl-ornithine hcl, dl-ornithine, hydrochloride, ornithine monohydrochloride, ornithine hydrochloride, ornithine, hydrochloride 1:1
hydrogen 2,5-diaminopentanoic acid chloride

Specifications

∼225°C (decomposition)
C5H13ClN2O2
4153338
Soluble in water.
[H+].[Cl-].NCCCC(N)C(O)=O
168.62
168.62
DL-Ornithine monohydrochloride
1069-31-4
MFCD00065398
dl-ornithine hydrochloride, 2,5-diaminopentanoic acid hydrochloride, dl-ornithine monohydrochloride, dl-ornithinehcl, h-dl-orn-oh.hcl, dl-ornithine hcl, dl-ornithine, hydrochloride, ornithine monohydrochloride, ornithine hydrochloride, ornithine, hydrochloride 1:1
GGTYBZJRPHEQDG-UHFFFAOYNA-N
hydrogen 2,5-diaminopentanoic acid chloride
71598
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-956-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only