accessibility menu, dialog, popup

UserName

CAS RN 1069-31-4

IUPAC Name: hydrogen 2,5-diaminopentanoic acid chloride
Synonyms: ornithine hcl
Molecular Weight (g/mol): 168.62
Molecular Formula: C5H13ClN2O2
InChi Key: GGTYBZJRPHEQDG-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NCCCC(N)C(O)=O
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (6)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)