Learn More
DL-Ornithine monohydrochloride, 99%
CAS: 1069-31-4 | C5H13ClN2O2 | 168.62 g/mol
Supplier: Thermo Scientific Chemicals A1817314
| Quantity | 25 g |
|---|
Description
Erythro-3-hydroxy-DL-ornithine monohydrochloride is used as a reagent to synthesize DL-Capreomycidine, a guanidine (HCl)-containing amino acid that is a component of antituberculous peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsErythro-3-hydroxy-DL-ornithine monohydrochloride is used as a reagent to synthesize DL-Capreomycidine, a guanidine (HCl)-containing amino acid that is a component of antituberculous peptides.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| C5H13ClN2O2 | |
| MFCD00065398 | |
| dl-ornithine hydrochloride, 2,5-diaminopentanoic acid hydrochloride, dl-ornithine monohydrochloride, dl-ornithinehcl, h-dl-orn-oh.hcl, dl-ornithine hcl, dl-ornithine, hydrochloride, ornithine monohydrochloride, ornithine hydrochloride, ornithine, hydrochloride 1:1 | |
| hydrogen 2,5-diaminopentanoic acid chloride |
Specifications
| ∼225°C (decomposition) | |
| 1069-31-4 | |
| MFCD00065398 | |
| dl-ornithine hydrochloride, 2,5-diaminopentanoic acid hydrochloride, dl-ornithine monohydrochloride, dl-ornithinehcl, h-dl-orn-oh.hcl, dl-ornithine hcl, dl-ornithine, hydrochloride, ornithine monohydrochloride, ornithine hydrochloride, ornithine, hydrochloride 1:1 | |
| GGTYBZJRPHEQDG-UHFFFAOYNA-N | |
| hydrogen 2,5-diaminopentanoic acid chloride | |
| 168.62 | |
| 168.62 |
| DL-Ornithine monohydrochloride | |
| C5H13ClN2O2 | |
| 4153338 | |
| Soluble in water. | |
| [H+].[Cl-].NCCCC(N)C(O)=O | |
| 25 g | |
| 71598 | |
| 99% |
Safety and Handling
EINECSNumber : 213-956-0
TSCA : Yes
Recommended Storage : Ambient temperatures