missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Norphenylephrine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 157°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 189.64 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | DL-Norphenylephrine Hydrochloride |
Safety and Handling
| RTECSNumber | DN7360000 |
| TSCA | No |