missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Methionine, FCC, 98.5-101.5%, Spectrum™ Chemical
ME126, 59-51-8, C5H11NO2S
$353.54 - $3,865.20
Chemical Identifiers
| CAS | 59-51-8 |
|---|---|
| Molecular Formula | C5H11NO2S |
| Molecular Weight (g/mol) | 149.21 |
| MDL Number | MFCD00063096 |
| InChI Key | FFEARJCKVFRZRR-UHFFFAOYNA-N |
| IUPAC Name | 2-amino-4-(methylsulfanyl)butanoic acid |
| SMILES | CSCCC(N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602300
|
Spectrum Chemical Mfg Cor
ME126500GM |
500 g | Amber Glass Bottle |
N/A
|
|
||||
|
18602301
|
Spectrum Chemical Mfg Cor
ME12625KG |
2.5 kg | Poly Pail |
N/A
|
|
||||
|
18602302
|
Spectrum Chemical Mfg Cor
ME12612KG |
12 kg | Fiber Drum |
N/A
|
|
||||
|
18602303
|
Spectrum Chemical Mfg Cor
ME12650KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical DL-Methionine, FCC is used as an amino acid nutritional supplement and flavor enhancer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 59-51-8 | |
| 149.21 | |
| FFEARJCKVFRZRR-UHFFFAOYNA-N | |
| CSCCC(N)C(O)=O |
| C5H11NO2S | |
| MFCD00063096 | |
| 2-amino-4-(methylsulfanyl)butanoic acid |
Specifications
| 59-51-8 | |
| 0.1% | |
| Amber Glass Bottle | |
| MFCD00063096 | |
| FFEARJCKVFRZRR-UHFFFAOYNA-N | |
| 2-amino-4-(methylsulfanyl)butanoic acid | |
| 98.5 to 101.5% |
| 100% | |
| 0.5% | |
| C5H11NO2S | |
| 500 g | |
| CSCCC(N)C(O)=O | |
| 149.21 | |
| FCC |