Learn More
DL-Methionine, FCC, 98.5-101.5%, Spectrum™ Chemical
ME126, 59-51-8, C5H11NO2S
Supplier:  Spectrum Chemical Mfg Cor ME12612KG
 View more versions of this product
Catalog No. 18602302
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical DL-Methionine, FCC is used as an amino acid nutritional supplement and flavor enhancer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
59-51-8
0.1%
Fiber Drum
MFCD00063096
FFEARJCKVFRZRR-UHFFFAOYNA-N
2-amino-4-(methylsulfanyl)butanoic acid
98.5 to 101.5%
100%
0.5%
C5H11NO2S
12 kg
CSCCC(N)C(O)=O
149.21
FCC
Chemical Identifiers
59-51-8
149.21
FFEARJCKVFRZRR-UHFFFAOYNA-N
CSCCC(N)C(O)=O
C5H11NO2S
MFCD00063096
2-amino-4-(methylsulfanyl)butanoic acid
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'