missing translation for 'onlineSavingsMsg'
Learn More

DL-Methionine, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18602303
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602303 50 kg Fiber Drum
18602300 500 g Amber Glass Bottle
18602301 2.5 kg Poly Pail
18602302 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602303 Supplier Spectrum Chemical Mfg Cor Supplier No. ME12650KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ME126, 59-51-8, C5H11NO2S

Spectrum™ Chemical DL-Methionine, FCC is used as an amino acid nutritional supplement and flavor enhancer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 59-51-8
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063096
InChI Key FFEARJCKVFRZRR-UHFFFAOYNA-N
IUPAC Name 2-amino-4-(methylsulfanyl)butanoic acid
SMILES CSCCC(N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Fiber Drum
Quantity 50 kg
Percent Purity 98.5 to 101.5%
Grade FCC