missing translation for 'onlineSavingsMsg'
Learn More

DL-Menthol, Crystal, USP, 98-102%, Spectrum™ Chemical

ME111, 89-78-1, C10H20O

$221.27 - $78,176.64

Chemical Identifiers

CAS 89-78-1
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
IUPAC Name (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18602294
View Documents Promotion Details
N/A 25 g Amber Glass Bottle
Each for $221.27
Only null left
 
18602295
View Documents Promotion Details
N/A 125 g Amber Glass Bottle
Each for $568.15
Only null left
 
18602296
View Documents Promotion Details
N/A 500 g Amber Glass Bottle
Each for $1,559.61
Only null left
 
18602298
View Documents Promotion Details
N/A 10 kg Amber Glass Bottle
Each for $9,027.36
Only null left
 
18602297
View Documents Promotion Details
N/A 50 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18602299
View Documents Promotion Details
N/A 180 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical DL-Menthol, Crystal, USP is used as a topical analgesic, antipruritic and decongestant. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

89-78-1
156.27
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
C10H20O
NOOLISFMXDJSKH-KXUCPTDWSA-N
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

Specifications

89-78-1
212°C
-2° to +2°
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
156.27
USP
DL-Menthol
100%
C10H20O
NOOLISFMXDJSKH-KXUCPTDWSA-N
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
98 to 102%
Crystals or Flakes
Videos
SDS
Documents

Documents