missing translation for 'onlineSavingsMsg'
Learn More

DL-Menthol, Crystal, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18602299
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
10 kg
50 kg
180 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602299 180 kg Amber Glass Bottle
18602294 25 g Amber Glass Bottle
18602295 125 g Amber Glass Bottle
18602296 500 g Amber Glass Bottle
18602298 10 kg Amber Glass Bottle
18602297 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18602299 Supplier Spectrum Chemical Mfg Cor Supplier No. ME111180KGBLK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ME111, 89-78-1, C10H20O

Spectrum™ Chemical DL-Menthol, Crystal, USP is used as a topical analgesic, antipruritic and decongestant. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 89-78-1
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
IUPAC Name (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

Specifications

CAS Max % 100%
Boiling Point 212°C
Packaging Amber Glass Bottle
Quantity 180 kg
Specific Rotation -2° to +2°
Percent Purity 98 to 102%
Grade USP
Physical Form Crystals or Flakes
Chemical Name or Material DL-Menthol