missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Menthol, Crystal, USP, 98-102%, Spectrum™ Chemical
ME111, 89-78-1, C10H20O
Supplier: Spectrum Chemical Mfg Cor ME11150KGBL
| Quantity | 50 kg |
|---|---|
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical DL-Menthol, Crystal, USP is used as a topical analgesic, antipruritic and decongestant. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 89-78-1 | |
| 156.27 | |
| (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol |
| C10H20O | |
| NOOLISFMXDJSKH-KXUCPTDWSA-N | |
| CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O |
Specifications
| DL-Menthol | |
| 100% | |
| Amber Glass Bottle | |
| 50 kg | |
| NOOLISFMXDJSKH-KXUCPTDWSA-N | |
| (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol | |
| 98 to 102% | |
| Crystals or Flakes |
| 89-78-1 | |
| 212°C | |
| C10H20O | |
| -2° to +2° | |
| CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O | |
| 156.27 | |
| USP |