missing translation for 'onlineSavingsMsg'
Learn More

DL-beta-Prolinol, 97+%, Thermo Scientific Chemicals

$541.44

Chemical Identifiers

CAS 5082-74-6
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD07773080
InChI Key QOTUIIJRVXKSJU-UHFFFAOYNA-N
Synonym 3-hydroxymethylpyrrolidine, 3-pyrrolidinemethanol, pyrrolidin-3-yl-methanol, pyrrolidine-3-yl-methanol, pyrrolidin-3-yl methanol, 3-pyrrolidinylmethanol, dl-beta-prolinol, rs-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine, 3-pyrrolidinemethanol, 3s
PubChem CID 13400657
IUPAC Name (pyrrolidin-3-yl)methanol
SMILES OCC1CCNC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH5722503
View Documents Promotion Details
N/A 1 g
Each for $541.44
Only null left
 
Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
Specifications

Chemical Identifiers

5082-74-6
101.15
QOTUIIJRVXKSJU-UHFFFAOYNA-N
13400657
OCC1CCNC1
C5H11NO
MFCD07773080
3-hydroxymethylpyrrolidine, 3-pyrrolidinemethanol, pyrrolidin-3-yl-methanol, pyrrolidine-3-yl-methanol, pyrrolidin-3-yl methanol, 3-pyrrolidinylmethanol, dl-beta-prolinol, rs-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine, 3-pyrrolidinemethanol, 3s
(pyrrolidin-3-yl)methanol

Specifications

5082-74-6
165°C to 171°C
C5H11NO
1420904
Slightly soluble in water
OCC1CCNC1
101.15
101.15
DL-beta-Prolinol
0.979
Hygroscopic
MFCD07773080
3-hydroxymethylpyrrolidine, 3-pyrrolidinemethanol, pyrrolidin-3-yl-methanol, pyrrolidine-3-yl-methanol, pyrrolidin-3-yl methanol, 3-pyrrolidinylmethanol, dl-beta-prolinol, rs-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine, 3-pyrrolidinemethanol, 3s
QOTUIIJRVXKSJU-UHFFFAOYNA-N
(pyrrolidin-3-yl)methanol
13400657
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

RUO – Research Use Only