missing translation for 'onlineSavingsMsg'
Learn More

DL-Adrenaline

Catalog No. p-7042574
Change view
Click to view available options
Quantity:
1 g
25 g
Catalog No. Quantity
AAJ6300503 1 g
AAJ6300514 25 g
2 options

Catalog No. AAJ6300503

Supplier: Thermo Scientific Chemicals J6300503

Only null left

A hormone and neurotransmitter | CAS: 329-65-7 | C9H13NO3 | 183.207 g/mol

A hormone and neurotransmitter

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 329-65-7
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00063027
InChI Key UCTWMZQNUQWSLP-UHFFFAOYSA-N
Synonym dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol
PubChem CID 838
ChEBI CHEBI:33568
IUPAC Name 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNCC(C1=CC(=C(C=C1)O)O)O

Specifications

Melting Point 197°C (decomposition)
Color White
Linear Formula (HO)2C6H3CH(CH2NHCH3)OH
Quantity 1 g
UN Number UN2811
Sensitivity Light sensitive
Merck Index 14,3619
Formula Weight 183.2
Physical Form Powder
Chemical Name or Material DL-Adrenaline
Hazard Category H311-H315-H319-H335
Hazard Statement GHS H Statement
H311-H315-H319-H335
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 206-347-6
RTECSNumber DO2975000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only