accessibility menu, dialog, popup

UserName

CAS RN 329-65-7

IUPAC Name: 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
Synonyms: (+-)-adrenaline racepinefrina dl-adrenaline epinephrine racemic racemic epinephrine racepinefrinum (+-)-epinephrine
Molecular Weight (g/mol): 183.21
Molecular Formula: C9H13NO3
InChi Key: UCTWMZQNUQWSLP-UHFFFAOYNA-N
SMILES: CNCC(O)C1=CC=C(O)C(O)=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)