missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Adrenaline 98.0+%, TCI America™
$87.60 - $561.08
Chemical Identifiers
| CAS | 329-65-7 |
|---|---|
| Molecular Formula | C9H13NO3 |
| Molecular Weight (g/mol) | 183.207 |
| MDL Number | MFCD00063027 |
| InChI Key | UCTWMZQNUQWSLP-UHFFFAOYSA-N |
| Synonym | dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol |
| PubChem CID | 838 |
| ChEBI | CHEBI:33568 |
| IUPAC Name | 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol |
| SMILES | CNCC(C1=CC(=C(C=C1)O)O)O |
Chemical Identifiers
| 329-65-7 | |
| 183.207 | |
| UCTWMZQNUQWSLP-UHFFFAOYSA-N | |
| 838 | |
| 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol |
| C9H13NO3 | |
| MFCD00063027 | |
| dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol | |
| CHEBI:33568 | |
| CNCC(C1=CC(=C(C=C1)O)O)O |
Specifications
| 329-65-7 | |
| C9H13NO3 | |
| 1 g | |
| dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol | |
| CNCC(C1=CC(=C(C=C1)O)O)O | |
| 183.207 | |
| CHEBI:33568 | |
| ≥98.0% (T) | |
| DL-Adrenaline |
| 197°C | |
| MFCD00063027 | |
| 2811 | |
| UCTWMZQNUQWSLP-UHFFFAOYSA-N | |
| 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol | |
| 838 | |
| 183.21 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0049
RTECSNumber : DO2975000
TSCA : No
Recommended Storage : Refrigerator