missing translation for 'onlineSavingsMsg'
Learn More

DL-Adrenaline 98.0+%, TCI America™

Catalog No. A01721G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01721G 1 g
A017225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A01721G Supplier TCI America Supplier No. A01721G

Chemical Identifiers

CAS 329-65-7
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00063027
InChI Key UCTWMZQNUQWSLP-UHFFFAOYSA-N
Synonym dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol
PubChem CID 838
ChEBI CHEBI:33568
IUPAC Name 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNCC(C1=CC(=C(C=C1)O)O)O

Specifications

Melting Point 197°C
Quantity 1 g
UN Number 2811
Formula Weight 183.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Adrenaline
EINECSNumber (9)-0049
RTECSNumber DO2975000
TSCA No
Recommended Storage Refrigerator