accessibility menu, dialog, popup

UserName

SOOOS

CAS RN 1043023-52-4

IUPAC Name: 8-oxa-3,13-dithiatricyclo[8.3.0.0²,⁶]trideca-1(10),2(6),4,11-tetraene-7,9-dione
Synonyms: 8-oxa-3,13-dithiatricyclo[8.3.0.0²,⁶]trideca-1(10),2(6),4,11-tetraene-7,9-dione
Molecular Weight (g/mol): 236.26
Molecular Formula: C10H4O3S2
InChi Key: UGFYLGZFYCTMFU-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C(SC=C2)C2=C1C=CS2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results