Learn More
Disuccinimidyl glutarate, 97%
CAS: 79642-50-5 | C13H14N2O8 | 326.26 g/mol
Supplier: Thermo Scientific Chemicals H58208ME
Description
Disuccinimidyl glutarate is used as a membrane-soluble, amine-reactive N-hydroxysuccinamide ester-based homobifunctional crosslinking reagent. It is a homobifunctionla cross-linking reagent, for ex. isolation of the insulin receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDisuccinimidyl glutarate is used as a membrane-soluble, amine-reactive N-hydroxysuccinamide ester-based homobifunctional crosslinking reagent. It is a homobifunctionla cross-linking reagent, for ex. isolation of the insulin receptor.
Solubility
Soluble in dimethylsulfoxide and dimethylformamide. Partially soluble in water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
| Disuccinimidyl glutarate | |
| C13H14N2O8 | |
| 500 mg | |
| Moisture sensitive | |
| Soluble in dimethylsulfoxide and dimethylformamide. Partially soluble in water. | |
| O=C(CCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O | |
| 326.26 | |
| 326.26 |
| 79642-50-5 | |
| MFCD00153597 | |
| 8575145 | |
| disuccinimidyl glutarate, di n-succinimidyl glutarate, bis 2,5-dioxopyrrolidin-1-yl pentanedioate, pentanedioic acid,1,5-bis 2,5-dioxo-1-pyrrolidinyl ester, dsg crosslinker, glutaric acid disuccinimidyl ester, di n-hydroxysuccinimidyl glutarate, bis 2,5-dioxopyrrolidin-1-yl glutarate, di n-succinimidyl glutarate chn | |
| LNQHREYHFRFJAU-UHFFFAOYSA-N | |
| bis(2,5-dioxopyrrolidin-1-yl) pentanedioate | |
| 4432628 | |
| 97% |
Chemical Identifiers
| 79642-50-5 | |
| 326.26 | |
| LNQHREYHFRFJAU-UHFFFAOYSA-N | |
| 4432628 | |
| O=C(CCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O |
| C13H14N2O8 | |
| MFCD00153597 | |
| disuccinimidyl glutarate, di n-succinimidyl glutarate, bis 2,5-dioxopyrrolidin-1-yl pentanedioate, pentanedioic acid,1,5-bis 2,5-dioxo-1-pyrrolidinyl ester, dsg crosslinker, glutaric acid disuccinimidyl ester, di n-hydroxysuccinimidyl glutarate, bis 2,5-dioxopyrrolidin-1-yl glutarate, di n-succinimidyl glutarate chn | |
| bis(2,5-dioxopyrrolidin-1-yl) pentanedioate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only